EMAIL THIS PAGE TO A FRIEND
GF30537569 Aldrich

Flax/Pp

(40% FLAX / PP), fabric, weight.m-2-420g, L 0.5 m, coil width 1250 mm

Synonym: FL323510

Purchase

Properties

form   fabric
mfr. no.   Goodfellow, 305-375-69
L   0.5 m
coil width   1250 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?