EMAIL THIS PAGE TO A FRIEND
GF38010192 Aldrich

Polystyrene

(PS), powder, mean particle size 900micron, weight 5000 g, condition expandable bead

Synonym: ST316090

Purchase

Properties

form   powder
mfr. no.   Goodfellow, 380-101-92
weight   5000 g

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2211 / PGIII

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?