• USA Home
  • GF40007887 - Polyphenylenesulfide - 40% Glass Fiber Reinforced

EMAIL THIS PAGE TO A FRIEND
GF40007887 Aldrich

Polyphenylenesulfide - 40% Glass Fiber Reinforced

(PPS - 40% GFR), granule, 3 mm nominal granule size, weight 2000 g

Synonym: SU316300

Purchase

Properties

mfr. no.   Goodfellow, 400-078-87
weight   2000 g
nominal granule size   3 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?