• USA Home
  • GF40889694 - Polyethylene Terephthalate

EMAIL THIS PAGE TO A FRIEND
GF40889694 Aldrich

Polyethylene Terephthalate

(polyester, PET, PETP), fabric, weight.m-2-92g, L 5 m, thickness 0.13 mm

Synonym: ES303520

Purchase

Properties

form   fabric
mfr. no.   Goodfellow, 408-896-94
L   5 m
thickness   0.13 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?