• USA Home
  • GF42784713 - Polymethylmethacrylate

EMAIL THIS PAGE TO A FRIEND
GF42784713 Aldrich

Polymethylmethacrylate

(PMMA, ACRYLIC), sheet, thickness 1.2 mm, size 200 × 200 mm, grade high purity

Synonym: ME303013

Purchase

Properties

form   sheet
mfr. no.   Goodfellow, 427-847-13
size   200 × 200 mm
thickness   1.2 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?