• USA Home
  • GF53170084 - Polyamide - Nylon 6, 6

EMAIL THIS PAGE TO A FRIEND
GF53170084 Aldrich

Polyamide - Nylon 6, 6

(PA 6,6), sheet, thickness 1 mm, size 500 × 500 mm

Synonym: AM323010

Purchase

Properties

form   sheet
mfr. no.   Goodfellow, 531-700-84
size   500 × 500 mm
thickness   1 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?