• USA Home
  • GF56898905 - Polyethylene - High Density

EMAIL THIS PAGE TO A FRIEND
GF56898905 Aldrich

Polyethylene - High Density

(HDPE), rod, diam. 100 mm, L 500 mm

Synonym: ET327980

Purchase

Properties

form   rod
mfr. no.   Goodfellow, 568-989-05
L   500 mm
diam.   100 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?