EMAIL THIS PAGE TO A FRIEND
GF74038901 Aldrich

Polystyrene

(PS), granule, 3.5 mm nominal granule size, weight 200 g, melt flow rate 2.5-3

Synonym: ST316310

Purchase

Properties

mfr. no.   Goodfellow, 740-389-01
weight   200 g
nominal granule size   3.5 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?