• USA Home
  • GF78949096 - Polyethersulfone

EMAIL THIS PAGE TO A FRIEND
GF78949096 Aldrich

Polyethersulfone

(PES), rod, diam. 20 mm, L 1000 mm, clear amber

Synonym: SU307920

Purchase

Properties

form   rod
mfr. no.   Goodfellow, 789-490-96
L   1000 mm
diam.   20 mm
color   clear amber

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?