• USA Home
  • GF90459928 - Polyamide - Nylon 6, 6

EMAIL THIS PAGE TO A FRIEND
GF90459928 Aldrich

Polyamide - Nylon 6, 6

(PA 6,6), granule, 8 mm nominal granule size, weight 2 kg, natural

Synonym: AM326330

Purchase

Properties

mfr. no.   Goodfellow, 904-599-28
weight   2 kg
color   natural
nominal granule size   8 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?