• USA Home
  • GF93858766 - Polyethersulfone

EMAIL THIS PAGE TO A FRIEND
GF93858766 Aldrich

Polyethersulfone

(PES), film, thickness 0.1 mm, size 500 × 500 mm, clear amber

Synonym: SU301100

Purchase

Properties

form   film
mfr. no.   Goodfellow, 938-587-66
size   500 × 500 mm
thickness   0.1 mm
color   clear amber

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?