• USA Home
  • GF94685866 - Polyethersulfone

EMAIL THIS PAGE TO A FRIEND
GF94685866 Aldrich

Polyethersulfone

(PES), rod, diam. 55 mm, L 100 mm, dark amber

Synonym: SU307955

Purchase

Properties

form   rod
mfr. no.   Goodfellow, 946-858-66
L   100 mm
diam.   55 mm
color   dark amber

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?