• USA Home
  • GF95250539 - Polyethylene - Medium Density

EMAIL THIS PAGE TO A FRIEND
GF95250539 Aldrich

Polyethylene - Medium Density

(MDPE), powder, max. particle size 350micron, weight 500 g

Synonym: EV306010

Purchase

Properties

form   powder
mfr. no.   Goodfellow, 952-505-39
weight   500 g

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?