EMAIL THIS PAGE TO A FRIEND
H1000 Aldrich

Heptadecanoic acid

98%

Synonym: C17:0, Heptadecylic acid, Margaric acid, NSC 3743

Purchase

Properties

InChI Key   KEMQGTRYUADPNZ-UHFFFAOYSA-N
assay   98%
bp   227 °C/100 mmHg(lit.)
mp   59-61 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MI3850000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

H3500 Heptadecanoic acid, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?