• USA Home
  • H5405 - 3-Hydroxy-7-methoxyflavone

EMAIL THIS PAGE TO A FRIEND
H5405 Aldrich

3-Hydroxy-7-methoxyflavone

Synonym: 7-Methoxyflavonol

Purchase

Properties

Related Categories Benzopyrans, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   IPRIGHIBTRMTDP-UHFFFAOYSA-N

Description

Packaging

250 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?