EMAIL THIS PAGE TO A FRIEND
H56509 Aldrich

4-Hydroxypteridine

98%

Synonym: 4-Pteridinol

  • CAS Number 700-47-0

  • Empirical Formula (Hill Notation) C6H4N4O

  • Molecular Weight 148.12

  •  MDL number MFCD00039725

  •  PubChem Substance ID 329815221

Purchase

Properties

InChI Key   UWWUXWWWOUNMKW-UHFFFAOYSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?