EMAIL THIS PAGE TO A FRIEND
L505 Aldrich

Lanthionine

96%

Synonym: Bis(2-Amino-2-carboxyethyl) sulfide, Bis(2-amino-2-carboxyethyl) sulfide

Purchase

Properties

InChI Key   DWPCPZJAHOETAG-UHFFFAOYSA-N
assay   96%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?