EMAIL THIS PAGE TO A FRIEND
L8300001-HH Aldrich

ETT Activator

0.25M, configured for Polygen, configured for PerkinElmer

Synonym: 5-(Ethylthio)-1H-tetrazole, 5-(Ethylthio)tetrazole solution

Purchase

Properties

Related Categories ABI 3900, ABI 39X, Activator, Chemical Synthesis, ETT Activator,
InChI Key   GONFBOIJNUKKST-UHFFFAOYSA-N
type   for DNA synthesis
product line   Proligo Reagents
assay   32-35 g/L (titration)
packaging   pkg of 6 × 200 mL L8300201-06
impurities   ≤30 ppm water content (Karl Fischer)
compatibility   configured for PerkinElmer
  configured for Polygen

Description

Preparation Note

Acetonitrile / 5-(Ethylthio)-1H-Tetrazole 100 / 3 v/w

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
3
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?