• USA Home
  • M199 - (±)-1-(3,4-Methylenedioxyphenyl)-2-butanamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
M199 Aldrich

(±)-1-(3,4-Methylenedioxyphenyl)-2-butanamine hydrochloride

>90%

Synonym: (±)-Benzodioxole-5-butanamine, BDB

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   LFRHMTZYADABJZ-UHFFFAOYSA-N
assay   >90%
drug control   USDEA Schedule I; Home Office Schedule 1; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
color   white
solubility   H2O: soluble

Description

Application

Analog of MDMA forensic reference standard.

Packaging

5 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?