• USA Home
  • M2502 - 7-Methylindole-3-carboxaldehyde

EMAIL THIS PAGE TO A FRIEND
M2502 Aldrich

7-Methylindole-3-carboxaldehyde

  • CAS Number 4771-50-0

  • Empirical Formula (Hill Notation) C10H9NO

  • Molecular Weight 159.18

  •  MDL number MFCD00047170

  •  PubChem Substance ID 24896736

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C10, C10-C12, Carbonyl Compounds,
InChI Key   KTUFZHVVJBHGKZ-UHFFFAOYSA-N
storage temp.   −20°C

Description

Application

Reactant for preparation of:
• Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
• Antibacterial and antituberculosis agents
• Antiandrogens effective against androgen receptor mutants
• Anti-bovine viral diarrhea virus (BVDV) agents
• Receptor subtype 5 antagonists
• Glucocorticoid receptor ligands
• Necroptosis inhibitors
• Orally efficacious small-molecule sctivator of the insulin receptor

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?