• USA Home
  • M2626 - (±)-m-Methoxymandelic acid

EMAIL THIS PAGE TO A FRIEND
M2626 Aldrich

(±)-m-Methoxymandelic acid

Synonym: DL-3-Methoxy-α-hydroxybenzeneacetic acid

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   YTRUJRPALUALSN-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?