EMAIL THIS PAGE TO A FRIEND
M87630 Aldrich

Morin hydrate

95%

Synonym: 2′,3,4′,5,7-Pentahydroxyflavone dihydrate, 2′,3,4′,5,7-Pentahydroxyflavone

  • CAS Number 654055-01-3

  • Empirical Formula (Hill Notation) C15H10O7 · xH2O

  • Molecular Weight 302.24 (anhydrous basis)

  •  MDL number MFCD00217054

  •  PubChem Substance ID 329818068

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   MYUBTSPIIFYCIU-UHFFFAOYSA-N
assay   95%
mp   299-300 °C (dec.)(lit.)

Description

Application

Reagent for the extraction and fluorometric determination of organotin compounds.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M4008 Morin hydrate, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?