Sign In to View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Skip To
assay
98%
bp
73-75 °C/10 mmHg (lit.)
mp
43-45 °C (lit.)
SMILES string
N#CC1CC2CCC1C2
InChI
1S/C8H11N/c9-5-8-4-6-1-2-7(8)3-6/h6-8H,1-4H2/t6-,7+,8?/m1/s1
InChI key
GAHKEUUHTHVKEA-KVARREAHSA-N
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
WGK 3
flash_point_f
174.2 °F - closed cup
flash_point_c
79 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
