• USA Home
  • N6898 - 5-Nitrothiophene-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
N6898 Aldrich

5-Nitrothiophene-2-carboxylic acid

≥98%

Purchase

Properties

Related Categories Building Blocks, C4 to C6, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   UNEPVPOHGXLUIR-UHFFFAOYSA-N
assay   ≥98%

Description

Packaging

10 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?