• USA Home
  • N7010 - 5-Nitro-2-n-propoxyaniline

EMAIL THIS PAGE TO A FRIEND
N7010 Aldrich

5-Nitro-2-n-propoxyaniline

Synonym: 1-n-Propoxy-2-amino-4-nitrobenzene, 2-Amino-4-nitro-1-n-propoxybenzene

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BY7400000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?