• USA Home
  • O6604 - tert-Octyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
O6604 Aldrich

tert-Octyl isothiocyanate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocyanates/Isothiocyanates More...
InChI Key   KUGHMFIBHMZICO-UHFFFAOYSA-N
assay   97%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?