• USA Home
  • O8755 - Oxalic acid dihydrate

EMAIL THIS PAGE TO A FRIEND
O8755 Aldrich

Oxalic acid dihydrate

97%

Synonym: Ethanedioic acid dihydrate

Purchase

Properties

vapor density   4.4 (vs air)
vapor pressure   <0.01 mmHg ( 20 °C)
InChI Key   GEVPUGOOGXGPIO-UHFFFAOYSA-N
assay   97%
mp   104-106 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

O0376 Oxalic acid dihydrate, ReagentPlus®, ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?