EMAIL THIS PAGE TO A FRIEND
P13622 Aldrich

2-Phenylethanol

99%

Synonym: β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA

Purchase

Properties

vapor density   4.21 (vs air)
vapor pressure   1 mmHg ( 58 °C)
InChI Key   WRMNZCZEMHIOCP-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.5317(lit.)
bp   219-221 °C/750 mmHg(lit.)
mp   −27 °C(lit.)
density   1.020 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
SG7175000
Flash Point(F) 
215.6 °F
Flash Point(C) 
102 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?