EMAIL THIS PAGE TO A FRIEND
P26759 Aldrich

Phenyl isocyanate

Purchase

Properties

vapor pressure   1.4 mmHg ( 20 °C)
InChI Key   DGTNSSLYPYDJGL-UHFFFAOYSA-N
refractive index   n20/D 1.535(lit.)
bp   162-163 °C(lit.)
mp   −30 °C(lit.)
density   1.096 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2487 3(6.1) / PGI
WGK Germany 
2
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?