• USA Home
  • P5657 - Poly(ethylene glycol) bis(6-aminohexyl)

EMAIL THIS PAGE TO A FRIEND
P5657 Aldrich

Poly(ethylene glycol) bis(6-aminohexyl)

Synonym: Polyoxyethylene bis(6-aminohexyl)

Purchase

Properties

storage temp.   2-8°C

Description

Preparation Note

Prepared from PEG with an average molecular weight of 3,350

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?