EMAIL THIS PAGE TO A FRIEND
P75202 Aldrich

L-Pyroglutamic acid

97%

Synonym: (S)-(−)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid

  • CAS Number 98-79-3

  • Empirical Formula (Hill Notation) C5H7NO3

  • Molecular Weight 129.11

  •  Beilstein Registry Number 82132

  •  EC Number 202-700-3

  •  MDL number MFCD00005272

  •  PubChem Substance ID 24898853

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Pyroglutamine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   ODHCTXKNWHHXJC-VKHMYHEASA-N
assay   97%
optical activity   [α]20/D −10°, c = 13 in H2O
mp   160-163 °C(lit.)

Description

Application

Building block for asymmetric synthesis and pharmaceuticals.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TW3710000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

83160 L-Pyroglutamic acid, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?