Synonym: PVP, Polyvidone, Povidone
CAS Number 9003-39-8
Linear Formula (C6H9NO)n
MDL number MFCD00149016
PubChem Substance ID 24899320
| mol wt | average mol wt 40,000 |
| viscosity number | 26-35(lit.) |
Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?