• USA Home
  • Q1883 - 3-Quinuclidinol hydrochloride

EMAIL THIS PAGE TO A FRIEND
Q1883 Aldrich

3-Quinuclidinol hydrochloride

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Quinuclidines
InChI Key   OYEJRVVBERZWPD-UHFFFAOYSA-N
assay   98%
mp   >300 °C(lit.)

Description

Packaging

10 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?