• USA Home
  • S4513 - Spermine diphosphate salt

EMAIL THIS PAGE TO A FRIEND
S4513 Aldrich

Spermine diphosphate salt

Synonym: N,N′-Bis(3-aminopropyl)-1,4-butanediamine tetrahydrochloride

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Organic Phosphates/Phosphites, Phosphorus Compounds More...
InChI Key   GOQWEECJRRZEHR-UHFFFAOYSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

Mixed NMDA agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?