-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
CAS Number 55-80-1
Linear Formula C15H17N3
Molecular Weight 239.323
MDL number MFCD00048014
Each unit = 50MG
Please note that Sigma-Aldrich does not collect analytical data for AldrichCPR products and that all sales are final.
All AldrichCPR products, and other building blocks or screening compounds that are part of this collection, are also available as custom packaged libraries - for more details about this service or for further information on this product, please contact us at AldrichCPR@sial.com.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?