• USA Home
  • SPC00022 - 3-(((Benzyloxy)carbonyl)amino)oxetane-3-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
SPC00022 Aldrich

3-(((Benzyloxy)carbonyl)amino)oxetane-3-carboxylic acid

Purchase

Properties

InChI Key   SDACJLQSZZYSTE-UHFFFAOYSA-N

Description

Other Notes

Product of SpiroChem

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?