• USA Home
  • SPC00023 - tert-Butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate

EMAIL THIS PAGE TO A FRIEND
SPC00023 Aldrich

tert-Butyl 3-(2-ethoxy-2-oxoethylidene)azetidine-1-carboxylate

Purchase

Properties

InChI Key   WYWJZDFQQJTRDD-UHFFFAOYSA-N

Description

Other Notes

Product of SpiroChem

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?