• USA Home
  • SPC00026 - tert-Butyl 3-(2-hydroxyethyl)azetidine-1-carboxylate

EMAIL THIS PAGE TO A FRIEND
SPC00026 Aldrich

tert-Butyl 3-(2-hydroxyethyl)azetidine-1-carboxylate

Purchase

Properties

InChI Key   PIEFOIGZJZFQQJ-UHFFFAOYSA-N

Description

Other Notes

Product of SpiroChem

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?