• USA Home
  • SPC00035 - 3-Fluorocyclobutanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
SPC00035 Aldrich

3-Fluorocyclobutanecarboxylic acid New

  • CAS Number 122665-96-7

  • Empirical Formula (Hill Notation) C5H7FO2

  • Molecular Weight 118.11

  •  MDL number MFCD09834117

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?