• USA Home
  • SPC00051 - 3-((Oxetan-3-ylamino)methyl)benzonitrile

EMAIL THIS PAGE TO A FRIEND
SPC00051 Aldrich

3-((Oxetan-3-ylamino)methyl)benzonitrile New

  • Empirical Formula (Hill Notation) C11H12N2O

  • Molecular Weight 188.23

  •  MDL number MFCD18849354

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?