• USA Home
  • SPC00064 - (3-(4-Chlorophenyl)oxetan-3-yl)methanamine

EMAIL THIS PAGE TO A FRIEND
SPC00064 Aldrich

(3-(4-Chlorophenyl)oxetan-3-yl)methanamine New

  • CAS Number 1260672-85-2

  • Empirical Formula (Hill Notation) C10H12ClNO

  • Molecular Weight 197.66

  •  MDL number MFCD18251063

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?