• USA Home
  • SPC00065 - (3-Phenyloxetan-3-yl)methanamine

EMAIL THIS PAGE TO A FRIEND
SPC00065 Aldrich

(3-Phenyloxetan-3-yl)methanamine New

  • CAS Number 497239-45-9

  • Empirical Formula (Hill Notation) C10H13NO

  • Molecular Weight 163.22

  •  MDL number MFCD18251065

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?