• USA Home
  • SPC00071 - trans-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
SPC00071 Aldrich

trans-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid New

  • CAS Number 939400-34-7

  • Empirical Formula (Hill Notation) C10H17NO4

  • Molecular Weight 215.25

  •  MDL number MFCD06657681

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?