• USA Home
  • SPC00078 - (3-Phenyloxetan-3-yl)methanol

EMAIL THIS PAGE TO A FRIEND
SPC00078 Aldrich

(3-Phenyloxetan-3-yl)methanol New

  • CAS Number 114012-43-0

  • Empirical Formula (Hill Notation) C10H12O2

  • Molecular Weight 164.20

  •  MDL number MFCD18642402

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?