• USA Home
  • SPC00089 - Cyclobutane-1,1-diylbis(methylene) bis(4-methylbenzenesulfonate)

EMAIL THIS PAGE TO A FRIEND
SPC00089 Aldrich

Cyclobutane-1,1-diylbis(methylene) bis(4-methylbenzenesulfonate) New

  • Empirical Formula (Hill Notation) C20H24O6S2

  • Molecular Weight 424.53

  •  MDL number MFCD22418996

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?