• USA Home
  • SPC00093 - 2,2′-(Cyclobutane-1,1-diyl)bis(ethan-1-ol)

EMAIL THIS PAGE TO A FRIEND
SPC00093 Aldrich

2,2′-(Cyclobutane-1,1-diyl)bis(ethan-1-ol) New

  • CAS Number 877125-96-7

  • Empirical Formula (Hill Notation) C8H16O2

  • Molecular Weight 144.21

  •  MDL number MFCD17012081

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?