• USA Home
  • SPC00096 - tert-Butyl 2,6-diazaspiro[3.4]octane-2-carboxylate

EMAIL THIS PAGE TO A FRIEND
SPC00096 Aldrich

tert-Butyl 2,6-diazaspiro[3.4]octane-2-carboxylate New

  • CAS Number 885270-84-8

  • Empirical Formula (Hill Notation) C11H20N2O2

  • Molecular Weight 212.29

  •  MDL number MFCD08234737

Purchase

Description

Legal Information

Product of SpiroChem.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?