EMAIL THIS PAGE TO A FRIEND
T7500 Aldrich

Tetrachloroethylene

ReagentPlus®, 99%

Synonym: PCE, Perchloroethylene

Purchase

Properties

Related Categories Cancer Research, Carcinogens, Cell Biology More...
grade   ReagentPlus®
vapor density   5.83 (vs air)
vapor pressure   13 mmHg ( 20 °C)
  19 mmHg ( 25 °C)
InChI Key   CYTYCFOTNPOANT-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.505(lit.)
bp   121 °C(lit.)
mp   −22 °C(lit.)
density   1.623 g/mL at 25 °C(lit.)

Description

Biochem/physiol Actions

Animal carcinogen that produces increased incidence of renal adenomas, adenocarcinomas, mononuclear cell leukemia, and hepatocellular tumors.

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1897 6.1 / PGIII
WGK Germany 
3
RTECS 
KX3850000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?