• USA Home
  • W218928 - Isobutyl isobutyrate

EMAIL THIS PAGE TO A FRIEND
W218928 Aldrich

Isobutyl isobutyrate

natural, ≥97%

Synonym: 2-Methylpropyl 2-methylpropanoate

Purchase

Properties

Related Categories Alphabetical Listings, Certified Natural Products, Flavors and Fragrances, I-L, I-N,
grade   Kosher
  natural
vapor pressure   1 mmHg ( 39.9 °C)
InChI Key   RXGUIWHIADMCFC-UHFFFAOYSA-N
assay   ≥97%
autoignition temp.   810 °F
refractive index   n20/D 1.398(lit.)
bp   145-152 °C(lit.)
mp   −81 °C(lit.)
density   0.855 g/mL at 25 °C(lit.)
Organoleptic   grape

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2528 3 / PGIII
WGK Germany 
2
RTECS 
NQ5250000
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles

Articles

The Complex Regulatory Landscape for Natural Flavor Ingredients

Introduction The Regulatory Landscape US and EU Definitions of Natural Regional Regulatory Variations The Example of Vanillin The Case for Standardization
Mr. Michael Sabisch1, Mr. David Smith2

1Senior Quality Assurance Specialist
2International Sales Coordinator, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food Safety, Oxidations

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?