• USA Home
  • W235504 - Cyclohexyl isovalerate

EMAIL THIS PAGE TO A FRIEND
W235504 Aldrich

Cyclohexyl isovalerate

≥98%

Synonym: Cyclohexyl 3-methylbutanoate

Purchase

Properties

Related Categories Alphabetical Listings, C-D, Flavors and Fragrances More...
biological source   synthetic
reg. compliance   FDA 21 CFR (172.515)
InChI Key   SQPOKBBCNZIWFI-UHFFFAOYSA-N
assay   ≥98%
refractive index   n20/D 1.441(lit.)
bp   223 °C(lit.)
density   0.925 g/mL at 25 °C(lit.)
Organoleptic   green; fruity; sweet

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?